Learn More
Medchemexpress LLC Acetylene-linker-Val-Cit-PABC-MMAE | 1411977-95-1 | 95.2% | 1307.62 g/mol | C67H106N10O16 | 10 MG

Supplier: Medchemexpress LLC HY1981210MG
Acetylene-linker-Val-Cit-PABC-MMAE is an antibody-drug conjugate (ADC) linker-payload conjugate that couples an alkyne-bearing cleavable Val-Cit-PABC linker to the tubulin inhibitor MMAE. The acetylene handle enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) for bioconjugation, while the Val-Cit-PABC motif permits proteolytic release of the payload after internalization.
- Contains an alkyne handle for CuAAC click chemistry
- Val-Cit-PABC motif enables proteolytic release of MMAE
- Provides a cleavable linker-payload architecture for ADC development
- Molecular formula C67H106N10O16 and molecular weight 1307.62 g/mol
- Typical purity approximately 95.2%
- Supplied as a 10 mg sample; store at 4°C under inert atmosphere
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.