Learn More
Strem, An Ascensus Company CAS# 1271-55-2. 100g. Acetylferrocene, 99.5%. MFCD00001432
SDP
Supplier:  Strem, An Ascensus Company 260050100G
Encompass
CAS# 1271-55-2. 100g. Acetylferrocene, 99.5%. MFCD00001432. Molecular Weight: 228.07. Molecular Formula: CH3COC5H4FeC5H5. Color/form: orange xtl. Strem# 26-0050. http://www.strem.com/catalog/v/26-0050/
Catalog No. 50-901-10942