Learn More
Cayman Chemical AcetylLcarnItInd3chLRI 10mg
SDP
Supplier:  Cayman Chemical 2656410 MG
Encompass_Preferred
An internal standard for the quantification of L-acetylcarnitine by GC- or LC-MS
Catalog No. 50-293-3848