Learn More
Sigma Aldrich Fine Chemicals Biosciences Acetylsalicylic acid, 50-78-2, MFCD00002430, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A5376100G
Encompass
Linear Formula: 2-(CH3CO2)C6H4CO2H, Molecular Weight: 180.16, ≥99.0%, mp: 134-136 °C, Synonym: 2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Aspirin.
Catalog No. NC0718758