Learn More
Medchemexpress LLC Acetylshikonin | 24502-78-1 | MFCD00143538 | 99.9% | 330.33 g/mol | C18H18O6 | 10 MG

Supplier: Medchemexpress LLC HYN218110MG
Acetylshikonin is a naphthoquinone derivative used in research for its anti-inflammatory, anticancer, antioxidant, antibacterial, and neuroprotective activities. It is supplied as a brown to khaki solid intended for laboratory research and analytical applications.
- High purity (approximately 99.9%).
- Brown to khaki solid appearance.
- Molecular formula C18H18O6 and molecular weight 330.33 g/mol.
- Available in small research pack sizes, including 10 MG.
- Store at 4°C and protect from light; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.