Learn More
Medchemexpress LLC HY-N2181 5mg Medchemexpress, Acetylshikonin CAS:24502-78-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21815MG
Encompass_Preferred
Medchemexpress, HY-N2181 5mg Acetylshikonin CAS:24502-78-1 Acetylshikonin, derived from the root of Lithospermum erythrorhizon, has anti-cancer and antiinflammation activity. Acetylshikonin is a non-selective cytochrome P450 inhibitor against all P450s (IC50 values range from 1.4-4.0 μM). Acetylshikonin is an AChE inhibitor and exhibits potent antiapoptosis activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2875