Learn More
Medchemexpress LLC Acetylthiocholine chloride | 6050-81-3 | MFCD00038727 | 98.0% | 197.73 g·mol⁻¹ | C7H16ClNOS | 250 MG

Supplier: Medchemexpress LLC HYW140953250MG
Acetylthiocholine chloride is a synthetic choline ester reagent used primarily as a substrate for acetylcholinesterase activity assays and other biochemical analyses. It is supplied as a solid powder suitable for laboratory assay workflows.
- Used as a substrate for acetylcholinesterase activity assays.
- Purity 98.0%.
- Molecular formula C7H16ClNOS; molecular weight 197.73 g·mol⁻¹.
- Available in multiple pack sizes, including 250 mg.
- Store sealed and protect from moisture; typical stability -80°C (6 months), -20°C (1 month, sealed).
- Supplied with supporting documentation such as a datasheet and certificate of analysis when available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.