Learn More
Selleck Chemical LLC Acetylthiocholine iodide E2998-50MG
SDP
Supplier:  Selleck Chemical LLC E299850MG
Encompass_Preferred
Acetylthiocholine iodide(S-Acetylthiocholine iodide ATCh iodide) is used as a substrate in the preparation of acetylcholine esterase (AChE) assay working solution for AChE activity assay In addition the compound is used in some medical research for example in the fields of neuroscience and organ physiology
Catalog No. 50-000-40438