Learn More
Sino Biological Assay Buffer and Co-factors: Acetyltransferase Dilution Buffer
Supplier:  Sino Biological A210920
Encompass_Preferred
Solution of buffer used for diluting enzyme histone acetyltransferases (KAT).
Catalog No. 50-256-9775