Learn More
Medchemexpress LLC AChE/BChE-IN-10 | 2924824-48-4 | C26H30N2O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY151368
Encompass_Preferred
AChE/BChE-IN-10 is a potent dual inhibitor of Acetylcholinesterase (AChE) and Butyrylcholinesterase (BChE), exhibiting good blood-brain barrier permeability. This compound is capable of inhibiting Aβ-aggregation and is utilized in Alzheimer's disease (AD) research.
  • Potent dual AChE and BChE inhibitor
  • Exhibits good blood-brain barrier permeability
  • Can inhibit Aβ-aggregation
  • Used in Alzheimer's disease (AD) research
Catalog No. 50-003-74450