Learn More
Medchemexpress LLC Achn-975 Tfa | 1410809-37-8 | 99.0% | 483.44 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY19936A
Encompass_Preferred
ACHN-975 TFA is a selective LpxC inhibitor that demonstrates subnanomolar LpxC inhibitory activity. It is effective against a broad spectrum of gram-negative bacteria, exhibiting low MIC values (≤1 μg/mL).
  • Selective LpxC inhibitor
  • Exhibits subnanomolar LpxC inhibitory activity
  • Effective against a wide range of gram-negative bacteria with low MIC values (≤1 μg/mL)
  • Potent against *P. aeruginosa* isolates
  • Demonstrates bactericidal activity against *P. aeruginosa* ATCC27853 strain in vivo.
Catalog No. 50-004-03054