Learn More
Medchemexpress LLC ACHP Hydrochloride | 406209-26-5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1306010MM/1ML
Encompass_Preferred
ACHP Hydrochloride (IKK-2 Inhibitor VIII) is a highly potent and selective IKK-β inhibitor with an IC50 of 8.5 nM. It is intended for research use only, targeting IKK and relevant to inflammation/immunology research areas. This substance is stable under recommended storage conditions.
  • Highly potent and selective IKK-β inhibitor
  • IC50 of 8.5 nM
  • Targets IKK
  • Identified for use as laboratory chemicals
  • Stable under recommended storage conditions
Catalog No. 50-003-39814