Learn More
Selleck Chemical LLC ACHP Hydrochloride-E1278-1MG
SDP
Supplier:  Selleck Chemical LLC E12781MG
Encompass_Preferred
ACHP Hydrochloride (IKK-2 Inhibitor VIII) is a highly potent and selective IKK- inhibitor with an IC50 of 8 5 nM
Catalog No. 50-313-8025