Learn More
Cayman Chemical AcIbenzolar-S-methyl 10mg
SDP
Supplier:  Cayman Chemical 2421210 MG
Encompass_Preferred
A fungicide inducer of systemic acquired resistance; induces resistance in wheat concomitant with expression of wheat chemically-induced (WCI) genes at 30 g per hectare; protective against V. inaequalis in apple trees when used in combination with light IPM
Catalog No. 50-293-3342