Learn More
Ethos Biosciences Acid Alcohol 0.5PCT 16oz
Supplier:  Ethos Biosciences 756016
Encompass_Preferred
Acid Alcohol 0 5% is a ready-to-use differentiating solution formulated for controlled decolorization in routine staining procedures Typically prepared as a dilute hydrochloric acid solution in alcohol it is used to selectively remove excess dye from tissue sections or cytology smears enhancing contrast and definition of stained structures Commonly applied in hematoxylin-based protocols and other staining methods requiring precise differentiation this solution supports consistent reproducible results Suitable for use in clinical research and educational laboratory environments
Catalog No. 50-008-19206