Learn More
Electron Microscopy Sciences Acid Alcohol 1% 500 ML
SDP
Supplier:  Electron Microscopy Sciences 2610203
Encompass_Preferred
For use in Truant's Fluorescent Method for Acid Fast - Organism For bacterial stain. See Truant, J.P. Henry Ford Hospital Med. Bull., 10:287, 1962. AFIP Manual of Histological Staining Methods, 3rd ed. Ed. L. Luna: New York: McGraw Hill Publications. c. 1968. p. 219. or Clark, G.: Staining Procedures. Williams and Wilkens Co., Baltimore. 3rd edition. c.1973. p. 225
Catalog No. 50-300-20