Learn More
Newcomer Supply Acid Alcohol 1%, 1 gallon
SDP
Supplier:  Newcomer Supply 10011C
Encompass
Use: H&E, AFB, PAS & Fite Stains. Shelf life is 2 years from date of manufacture. Store at 15-30°C
Catalog No. NC9451315