Learn More
Medchemexpress LLC Acid-C2-PEG4-C2-NHS ester | 1343476-41-4 | 98.0% | 50 MG

Supplier: Medchemexpress LLC HY13043650MG
Acid-C2-PEG4-C2-NHS ester is a PEG-based PROTAC linker. It is utilized in the synthesis of PROTACs, which consist of two distinct ligands connected by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. These PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
- Molecular weight: 435.42
- Molecular formula: C18H29NO11
- Appears as a colorless to light yellow liquid
- Density: 1.30±0.1 g/cm3
- Soluble in DMSO at 100 mg/mL
- For research use only
- Available documentation includes data sheet, SDS, COA, HNMR, and MS
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.