Learn More
Cambridge Isotope Laboratories ACID CAL FOR QNMR 5MM (160UL)
Supplier:  Cambridge Isotope Laboratories DLM9491B
Encompass
ACID CALIBRANT FOR QNMR 5mM BENZOIC ACID IN DMSO-D6 (160UL FILL VOLUME)
Catalog No. NC1885261