Learn More
Medchemexpress LLC Acid-PEG2-C2-Boc | 2086688-99-3 | ≥98.0% | C12H22O6 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140480
Encompass_Preferred
Acid-PEG2-C2-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Contains ligands for E3 ubiquitin ligase and target protein
Catalog No. 50-003-52884