Learn More
Medchemexpress LLC HY-140112 100mg Medchemexpress, Acid-PEG2-SS-PEG2-acid CAS:1807539-10-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY140112100MG
Encompass_Preferred
Medchemexpress, HY-140112 100mg Acid-PEG2-SS-PEG2-acid CAS:1807539-10-1 Acid-PEG2-SS-PEG2-acid is a cleavable 4 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1549