Learn More
Medchemexpress LLC Acid-PEG3-mono-methyl ester | 1807505-26-5 | ≥98.0% | C11H20O7 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY140484
Encompass_Preferred
Acid-PEG3-mono-methyl ester is an alkyl/ether-based PROTAC linker. It can be used in the synthesis of PROTACs, which are designed with two different ligands connected by a linker. One ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs work by leveraging the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-52719