Learn More
Medchemexpress LLC Acid-PEG3-SS-PEG3-acid | 2055014-98-5 | 98.0% | 474.59 g/mol | C18H34O10S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY14011325MG
Encompass_Preferred
Acid-PEG3-SS-PEG3-acid is a cleavable six-unit polyethylene glycol (PEG) linker used in the synthesis of antibody-drug conjugates (ADCs). The molecule has carboxylic acid termini that enable amide-coupling bioconjugation and a disulfide (SS) linkage that allows intracellular cleavage under reducing conditions.
  • Cleavable six-unit PEG linker
  • Carboxylic acid termini for amide coupling
  • Disulfide (SS) cleavable linkage for intracellular release
  • Supplied in multiple pack sizes for research use
Catalog No. 50-002-04068