Learn More
Medchemexpress LLC Acid Red 249 | 6416-66-6 | 95.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYD04301G
Encompass_Preferred
Acid Red 249, also known as Tracid Brilliant Red B, is a weak acid dye characterized by its sulfate ion content. It presents as a solid, typically brown to reddish-brown in color, and has a molecular weight of 748.11 with the formula C29H20ClN3Na2O10S3. This product is primarily used as a fluorescent dye in various research applications.
  • Functions as a fluorescent dye.
  • Weak acid dye containing sulfate ion.
  • Solid appearance, brown to reddish-brown color.
  • Molecular weight of 748.11.
Catalog No. 50-004-10394