Learn More
ATCC Pfizer Pneumococcal; Polysaccharide powder; Type 64 (23B) 10mg
Supplier:  ATCC 549X
Encompass
Purified pneumococcal polysaccharide type 23B is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 23B can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-0241
 Restricted Item