Learn More
Medchemexpress LLC Acifluorfen | 50594-66-6 | 98.95% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY128075100MG
Encompass_Preferred
Acifluorfen is a protoporphyrinogen oxidase (PROTOX) inhibitor herbicide that promotes the accumulation of protoporphyrin IX (PPIX) and induces tumors in the rodent liver. It also causes strong photooxidative destruction of pigments and lipids in sensitive plant species.
  • Promotes accumulation of protoporphyrin IX (PPIX)
  • Induces tumors in rodent liver
  • Causes strong photooxidative destruction of pigments and lipids in sensitive plant species
Catalog No. 50-003-38888