Learn More
Medchemexpress LLC Acifran | 72420-38-3 | MFCD00865746 | 99.3% | 218.21 g·mol^-1 | C12H10O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10757925MG
Encompass_Preferred
Acifran (AY 25712) is an orally active antihyperlipidemic agent that acts as an agonist of GPR109A (HM74A) and GPR109B, the high- and low-affinity receptors for niacin. It is provided as a research-grade compound for pharmacological and biochemical studies; users should consult the supplier datasheet for handling, purity, and storage details.
  • Orally active agonist of GPR109A and GPR109B.
  • Suitable for studying niacin receptor signaling and lipid metabolism.
  • Supplied in small milligram quantities for research use.
  • Stable as a powder at -20°C; solvent storage conditions documented.
  • High purity appropriate for analytical and biological assays.
Catalog No. 50-002-41575