Learn More
Apexbio Technology LLC Acitretin 55079-83-9 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A24155.1
Encompass_Preferred
Acitretin (CAS 55079-83-9) is a synthetic aromatic retinoid acting as an agonist of nuclear retinoic acid receptors (RARs) It is designed to modulate gene transcription thereby influencing cellular differentiation and proliferation Acitretin exerts its biological activity primarily through binding and activating RARs affecting retinoid signaling pathways involved in keratinocyte differentiation and epidermal proliferation Based on these pharmacological properties Acitretin holds research potential in the study of psoriasis other dermatological disorders and Alzheimer s disease
Catalog No. 50-000-95673