Learn More
Cayman Chemical AcLEHDAFCtrIfluoroACTAte 10mg
SDP
Supplier:  Cayman Chemical 1705110 MG
Encompass_Preferred
A fluorogenic substrate that can be cleaved by caspase-4, -5, and -9
Catalog No. 50-296-6603