Learn More
TARGETMOL CHEMICALS INC ACLIDINIUM BROMIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T255525MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Aclidinium bromide (LAS-W 330) is a synthetic anticholinergic agent that is used as an inhalant for treatment of acute bronchospasm due to chronic bronchitis or emphysema. Aclidinium has not been implicated in causing liver enzyme elevations or clinically apparent acute liver injury. purity: 98%
Catalog No. 50-258-0745