Learn More
Sino Biological AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 Antibody, Mouse MAb
Supplier:  Sino Biological 40496MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant AcmNPV Envelope glycoprotein gp64 / AcmNPV-gp64 ( Catalog#40496-V08B; NP_054158.1; Met1-Thr481). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-3713