Learn More
Medchemexpress LLC Acoltremon | 68489-09-8 | 99.1% | 289.41 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY108449500MG
Encompass_Preferred
Acoltremon is a potent and selective TRPM8 agonist, which is a menthol derivative that acts as a cooling agent. This compound demonstrates an analgesic effect and can be utilized in chronic neuropathic pain research.
  • Acts as a potent and selective TRPM8 agonist.
  • Functions as a cooling agent.
  • Shows analgesic effect.
  • Can be used in chronic neuropathic pain research.
  • Displays TRPM8 agonism in tests on heterologously expressed TRP ion channels in vitro.
  • Exhibits highly potent and specific TRPM8 channel agonist activity in DRG neurons in vitro.
  • Activates mouse TRPM8 but does not activate or inhibit mouse TRPA1 in HEK293T cells.
  • Produces an obvious analgesic effect in mice via selective TRPM8 activation.
  • Reduces capsaicin-induced nocifensive behavior in mice.
Catalog No. 50-003-45480