Learn More
Cayman Chemical AconItase FRrous AmonIM Su 1ea
SDP
Supplier:  Cayman Chemical 7055081 EA
Encompass_Preferred
An a1-adrenergic and 5-HT2A receptor antagonist (IC50s 2A- and a2B-adrenergic and 5-HT1A 5-HT1B 5-HT2B and 5-HT2C receptors (IC50s 18.1 4.89 4.68 41 11.5 and 26 nM respectively) among others decreases systolic and diastolic blood pressure and heart rate in spontaneously hypertensive rats and 2K-1C renal hypertensive dogs at 10 and 4 mg/kg respectively
Catalog No. 50-289-3259