Learn More
Cayman Chemical Acontum carmchal Dbx Acn 10mg
SDP
Supplier:  Cayman Chemical 2504810 MG
Encompass_Preferred
An active metabolite of aconitine; inhibits osteoclast differentiation of RANKL-stimulated RAW 264.7 cells and bone resorption in a pit formation assay; inhibits RANKL-induced activation of NF-κB and NFATc1 in RAW 264.7 cells; toxic to mice at 120 mg/kg; induces flaccid paralysis and toxicity in rats (TD50 = 1.5 μmol/animal; LD50 = 1.7 μmol/animal)
Catalog No. 50-293-3748