Learn More
Medchemexpress LLC Acoramidis hydrochloride | 2242751-53-5 | 98.7% | 328.77 | 1 MG
Supplier: Medchemexpress LLC HY109165A1MG
Acoramidis (AG10) hydrochloride is an orally active and selective kinetic stabilizer of both wild-type (WT) and V122I-TTR (transthyretin). It is utilized in studies related to transthyretin amyloidosis.
- Orally active.
- Selective kinetic stabilizer of WT and V122I-TTR.
- Stabilizes V122I- and WT-TTR equally well and exceeds tafamidis in stabilizing WT and mutant TTR in whole serum at concentrations between 0.1-10 μM.
- Stimulates mitochondrial QO2 in a concentration-dependent manner between 10 and 100 μM.
- Exhibits minimal inhibition of common off-targets, such as the potassium ion channel hERG (IC50 > 100 μM) and cytochrome P450 isozymes (IC50 > 50 μM), indicating low toxicity.
- In Wistar rats, an oral gavage of 50 mg/kg/d for 28 days showed a plasma Cmax of ~40 μM with no pathological processes in liver, kidney, heart, spleen, thymus, and lung.