Learn More
Selleck Chemical LLC Acotiamide hydrochloride S4838-25mg
SDP
Supplier:  Selleck Chemical LLC S483825MG
Encompass_Preferred
Acotiamide Hydrochloride (YM-443 Z-338) is the hydrochloride salt form of acotiamide a prokinetic agent with gastrointestinal (GI) motility-enhancing activity It is a new orally active selective acetylcholinesterase inhibitor
Catalog No. 50-311-1910