Learn More
Medchemexpress LLC ACP-105 | 899821-23-9 | 98.9% | 290.79 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY11225610MM/1ML
Encompass_Preferred
ACP-105 is an orally available, selective and potent androgen receptor modulator (SARM), with pEC50s of 9.0 and 9.3 for AR wild type and T877A mutant, respectively.
  • Orally available
  • Selective androgen receptor modulator (SARM)
  • Potent activity against AR wild type and T877A mutant
Catalog No. 50-004-35881