Learn More
Medchemexpress LLC Acp-5862 | 2230757-47-6 | 98.3% | C26H23N7O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13533410MG
Encompass_Preferred
ACP-5862 is the major active, circulating, pyrrolidine ring-opened metabolite of acalabrutinib that inhibits Bruton tyrosine kinase (BTK). It has been characterized for its contribution to acalabrutinib's pharmacological activity and displays potent BTK inhibition alongside weak, time-dependent inactivation of select cytochrome P450 enzymes. Supplied for research use with supporting analytical documentation.
  • Potent BTK inhibition (IC50 ≈ 5.0 nM).
  • Major circulating metabolite of acalabrutinib.
  • Weak time-dependent inactivator of CYP3A4 and CYP2C8.
  • Available as solid and as a DMSO solution.
  • High analytical purity (~98.3% by HPLC).
  • Supplied with certificate of analysis and safety data sheet.
Catalog No. 50-002-58586