Learn More
Waters Corp Waters ACQUITY APC AQ Column, 900 Å, 2.5 μm, 4.6 x 30 mm
Supplier:  Waters Corp 186007249
Encompass_Preferred
ACQUITY™ Advanced Polymer Chromatography™ (APC™) Columns feature high-performance chemistries for rapid, accurate chromatographic characterization of synthetic polymers and macromolecules. Innovative hybrid-polymer sub-3-μm particle technology offers unmatched chromatographic efficiency, stability, and method flexibility for low molecular weight aqueous and organic soluble polymers up to 2,000,000 g/mole.
  • Rapid, accurate characterization of synthetic polymers and macromolecules.
  • Hybrid-polymer sub-3-μm particle technology for efficiency and stability.
  • Suited for low molecular weight aqueous/organic soluble polymers up to 2,000,000 g/mole.
  • Designed for aqueous-based separations.
  • Employs BEH Technology for extreme performance and long column lifetime.
  • Rigid BEH particle prevents shrinking and swelling.
Catalog No. 50-239-1394