Learn More
Waters Corp Waters ACQUITY APC AQ BEH Column, 900Å, 2.5 μm
Supplier:  Waters Corp 186007250
Encompass_Preferred
ACQUITY APC AQ columns are designed to provide highly efficient size-based separations for water-soluble polymer characterization.
  • Sorbent utilizes patented Ethylene Bridge Hybrid (BEH) technology.
  • Offers a mechanically strong packed bed that resists shrinking and swelling.
  • Facilitates easy mobile phase solvent switching.
  • Expected molecular weight range is approximately 300,000 to 2,000,000 g/mole.
Catalog No. NC2313319