Learn More
Waters Corp Calibration Kit, Polystyrene, High Molecular Weight, Gel Permeation
Supplier:  Waters Corp 186007541
Encompass_Preferred
This calibration kit provides pre-prepared polystyrene standards specifically designed for molecular weight analysis with ACQUITY APC XT Columns. It eliminates manual preparation of custom mixes, offering an efficient and ready-to-use solution for targeted molecular weight range calibration.
  • Designed for molecular weight analysis with ACQUITY APC XT Columns.
  • Eliminates manual preparation of custom standard calibration mixes.
  • Provides appropriate data points for the targeted molecular weight range.
  • Packaged in 12 x 32 mm clear glass vials with PTFE/silicone septa.
  • Cap colors (yellow, purple, pink) indicate specific molecular weight standards.
  • Ready-to-use: simply add solvent to dissolve standards and place in autosampler.
  • Contains multiple vials with specific polystyrene standards, including Mp 1800K, 1210K, 552K, and others, spanning a high molecular weight range.
Catalog No. 50-239-1339