Learn More
Waters Corp ACQUITY APC XT BEH Column, 200 Å, 2.5 μm, 4.6 x 75 mm
Supplier:  Waters Corp 186007003
Encompass_Preferred
ACQUITY APC XT columns provide highly efficient size-based separations for organic solvent soluble polymer characterization. Patented Ethylene Bridged Hybrid (BEH) technology ensures a mechanically strong packed bed, resisting shrinking and swelling for easy mobile phase solvent switching. These columns offer superior efficiency, stability, and method flexibility for synthetic polymer analysis.
  • Optimized pore volume and particle size for unmatched separation speed, resolution, and efficiency.
  • Rigid BEH particle provides stable bed, preventing shrinking/swelling during rapid solvent switching.
  • Tightly controlled manufacturing ensures consistent product reproducibility.
  • Intelligent Chip Technology tracks column performance and usage history.
  • BEH technology particle synthesis from TEOS and BTEE for a highly stable and mechanically strong particle.
Catalog No. 50-239-0784