Learn More
Waters Corp Waters Method Validation Kit, Glycan, 1.7 μm, 2.1 mm x 100 mm
Catalog No. 50466125 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit for glycan analysis utilizes Hydrophilic Interaction Liquid Chromatography (HILIC) with UPLC technology. It contains three batches of chromatographic media to assess method quality. Columns feature MaxPeak High-Performance Surface technology and support a broad pH range for separating glycans.
  • Exceptional batch-to-batch and column reproducibility.
  • Includes three batches of chromatographic media for method quality assessment.
  • QC tested with 2-AB labeled glycans for consistent performance.
  • MaxPeak High-Performance Surface technology prevents unwanted analyte-surface interactions.
  • Offers a wide, usable pH range for separating neutral and acidic glycans.
  • Ethylene-Bridged Hybrid (BEH) base particle ensures chemical and mechanical stability.
  • Designed to withstand increased UPLC flow rates and backpressures.
Catalog No. 50-466-125 Supplier Waters Corp Supplier No. 186004907
This method validation kit for glycan analysis utilizes Hydrophilic Interaction Liquid Chromatography (HILIC) with UPLC technology. It contains three batches of chromatographic media to assess method quality. Columns feature MaxPeak High-Performance Surface technology and support a broad pH range for separating glycans.
  • Exceptional batch-to-batch and column reproducibility.
  • Includes three batches of chromatographic media for method quality assessment.
  • QC tested with 2-AB labeled glycans for consistent performance.
  • MaxPeak High-Performance Surface technology prevents unwanted analyte-surface interactions.
  • Offers a wide, usable pH range for separating neutral and acidic glycans.
  • Ethylene-Bridged Hybrid (BEH) base particle ensures chemical and mechanical stability.
  • Designed to withstand increased UPLC flow rates and backpressures.