Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters BEH HILIC VanGuard Pre-column, 1.7 μm, 130Å

Supplier:  Waters Corp 186003980

Encompass_Preferred

This VanGuard Pre-column protects analytical columns by preventing chemical and particulate impurities from entering the mobile phase stream. Designed for direct attachment to UPLC columns, it extends column lifetime, maintains separation efficiency, minimizes extra column volumes, and reduces leaks.

  • Directly attaches to UPLC columns, minimizing extra column volumes and preventing leaks.
  • Enhances analytical performance by preventing chemical and particulate impurities.
  • Ensures long analytical column lifetime and maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Manufactured under strict cGMP, ISO 9001 conditions, tested and certified for optimal performance.
  • Tolerates pressures up to 18000 psi (1240 bar).
  • Features BEH Technology, 130Å pore, and 1.7μm particles.
  • Each batch is QC tested with a peptide map for consistent performance.

Catalog No. 50-782-107


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.