Learn More
Waters Corp Waters Column, C18, 1.7 μm, 3 mm X 50 mm, 130 Å
Supplier:  Waters Corp 186005300
Encompass_Preferred
ACQUITY CSH C18 columns provide alternate selectivity for reversed-phase UPLC. They address peak shape asymmetry, poor loading capacity for basic compounds, column bleed, and mobile-phase re-equilibration issues. These columns deliver broad selectivity and high performance, including superior peak shape, low column bleed, and high efficiency.
  • Provides TFA peak shape performance without TFA drawbacks.
  • Improved peak performance with MS-friendly mobile phases like formic acid.
  • Faster, easier method development as formic acid lacks lingering chromatographic effects.
  • Offers largest loading of basic chemicals under volatile, acidic, low pH mobile-phase conditions, reducing solvent usage and fraction volumes.
  • Consistent results and reduced replacement need due to strict manufacturing.
Catalog No. 50-791-330