Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Column, 1.7 μm, 130 Å
Supplier:  Waters Corp 176002139
Encompass_Preferred
ACQUITY CSH C18 columns are ultra-efficient UPLC columns offering alternate selectivity for reversed-phase separations. Featuring Charged Surface Hybrid (CSH) Technology and UPLC integration, they enhance peak shape, loading capacity, resolution, and analysis speed.
  • Provides distinct separation characteristics for reversed-phase UPLC.
  • CSH technology enhances selectivity, peak shape, and loading capacity for basic compounds.
  • UPLC technology improves resolution, sensitivity, and analysis speed with HPLC method robustness.
  • Individually tested with a performance chromatogram and certificate of batch analysis.
Catalog No. 50-791-108