Learn More
Waters Corp Waters ACQUITY UPLC CSH C18 Method Validation Kit, 1.7 μm, 3 mm × 100 mm, 3/pk
Supplier: Waters Corp 186005574
This kit consists of three ACQUITY CSH C18 columns packed with three different batches of sorbent.
- C18 chemistry
- Reversed phase separation mode
- Hybrid particle substrate
- pH range of 1 to 11
- Maximum pressure of 18000 psi (1240 Bar)
- Endcapped
- Low silanol activity
- Spherical particle shape
- 1.7 μm particle size
- Parker-style endfitting type
- 130 Å pore size
- Method validation kit format
- Surface area of 185
- Compatible with UPLC and UHPLC systems
- CSH particle technology
- L1 USP classification
- 3 mm inner diameter
- 100 mm length
- 15% carbon load
- eCord enabled
- 3 units per package