Learn More
Waters Corp WATERS CORPORATION Column, 1.7 μm, 3 mm x 50 mm, Fluoro-Phenyl
Supplier:  Waters Corp 186005355
Encompass_Preferred
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column offers maximum method development freedom, particularly when using low pH mobile phases. The non-encapped sorbent provides enhanced retention of acidic compounds.
  • Maximizes selectivity differences.
  • Provides maximum method development freedom, especially with low pH mobile phases.
  • Non-encapped sorbent enhances retention of acidic compounds.
Catalog No. 50-791-375