Learn More
Waters Corp Waters ACQUITY UPLC CSH Phenyl-Hexyl Column, 130 Å, 1.7 μm, 3 mm x 150 mm
Supplier:  Waters Corp 186005412
Encompass_Preferred
ACQUITY CSH Phenyl-Hexyl columns offer alternative analyte selectivity, ideal for method development. They feature a trifunctionally bonded C6 phenyl ligand, a robust and low-bleed sorbent, selectively retaining polyaromatic compounds through π–π interactions.
  • Alternative analyte selectivity
  • Valuable tool for method development
  • Robust, low bleed sorbent
  • Selectively retains polyaromatic compounds via π–π interactions
  • Powered by charged surface hybrid (CSH) particle technology
  • Superior peak shape across low and high pH conditions
  • Complementary selectivity to straight-chain alkyl phases
  • Optimized loading capacity for basic compounds at both low and high pH
  • Eliminates ion-pair reagents
  • Exceptional stability for advanced column equilibration
  • Highly reproducible through chromatographic testing
Catalog No. 50-791-425