Learn More
Waters Corp Waters Corporation Column, 1.7 μm, 2.1 mm ID, 50 mm, Phenyl-Hexyl
Supplier:  Waters Corp 186005406
Encompass_Preferred
These columns offer alternative analyte selectivity for method development. The trifunctionally bonded C6 phenyl ligand provides robust, low bleed performance, retaining polyaromatic compounds via pi-pi interactions. Built on the Charged Surface Hybrid (CSH) particle platform, they ensure complementary selectivity and exceptional peak shape under varying pH conditions.
  • Robust, low bleed performance with selective retention of polyaromatic compounds.
  • CSH particle platform offers complementary selectivity and exceptional peak shape.
  • High efficiency, superior loading capacity, and improved reproducibility.
  • Manufactured in a cGMP, ISO 9001:2000 certified plant, with each batch tested for performance.
Catalog No. 50-791-421